2,4-Difluoroaniline 97%

Supplier: Ambeed
Danger

A518706-10G A518706-25G A518706-5G A518706-100G
77103-196EA 6 USD
77103-196 77103-198 77103-200 77613-630
2,4-Difluoroaniline 97%
2,4-Difluoroaniline

Precaución: for Research Use Only

Formula: C₆H₅F₂N
MDL Number: MFCD00007648
CAS Number: 367-25-9
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR