2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 4-Formyl-3-methoxyphenylboronic acid pinacol ester

A515862-1G A515862-250MG A515862-5G
77103-166EA 109.28 USD
77103-166 77103-168 77103-170
2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 98%
2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₉BO₄
MW: 262.10 g/mol
MDL Number: MFCD18732927
CAS Number: 956431-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR