2,3-Dimethylbenzylamine 98%

Supplier: Ambeed
Warning

A515575-1G A515575-25G A515575-5G
77103-154EA 16 USD
77103-154 77103-156 77103-158
2,3-Dimethylbenzylamine 98%
2,3-Dimethylbenzylamine

Precaución: for Research Use Only

Formula: C₉H₁₃N
MDL Number: MFCD00052680
CAS Number: 51586-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR