5-Amino-2-(trifluoromethoxy)benzoic acid 98%

Supplier: Ambeed
Warning

A514587-1G A514587-250MG
77103-148EA 81.69 USD
77103-148 77103-150
5-Amino-2-(trifluoromethoxy)benzoic acid 98%
5-Amino-2-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₃
MW: 221.13 g/mol
MDL Number: MFCD15527344
CAS Number: 307989-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR