2-(2-Bromo-5-methylphenyl)acetic acid 95%

Supplier: Ambeed
Warning

A514172-100MG A514172-1G A514172-250MG A514172-5G
77103-134EA 26 USD
77103-134 77103-136 77103-138 77103-140
2-(2-Bromo-5-methylphenyl)acetic acid 95%
2-(2-Bromo-5-methylphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MDL Number: MFCD02664810
CAS Number: 203314-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR