4-Butyrylbiphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 4'-Phenylbutyrophenone

A512378-100G A512378-25G A512378-500G A512378-5G
77103-120EA 26.53 USD
77103-120 77103-122 77103-124 77103-126
4-Butyrylbiphenyl 98%
4-Butyrylbiphenyl

Precaución: for Research Use Only

Formula: C₁₆H₁₆O
MW: 224.30 g/mol
MDL Number: MFCD00022005
CAS Number: 13211-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR