3-Anilino-1-propanol 95%

Supplier: Ambeed
Warning

Synonyms: N-(3-Hydroxypropyl)aniline, 3-(Phenylamino)propan-1-ol

A512038-10G A512038-1G A512038-250MG
77103-114EA 319.34 USD
77103-114 77103-116 77103-118
3-Anilino-1-propanol 95%
3-Anilino-1-propanol

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MW: 151.21 g/mol
MDL Number: MFCD15206074
CAS Number: 31121-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR