2-Bromo-6-hydroxy-4-methoxybenzaldehyde 97+%

Supplier: Ambeed
Warning

A512017-100MG A512017-1G A512017-250MG
77103-108EA 37.14 USD
77103-108 77103-110 77103-112
2-Bromo-6-hydroxy-4-methoxybenzaldehyde 97+%
2-Bromo-6-hydroxy-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
MW: 231.043 g/mol
MDL Number: MFCD23705610
CAS Number: 861668-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR