2-Bromo-5-hydroxymethylaniline 97%

Supplier: Ambeed
Warning

A511779-1G A511779-250MG A511779-5G
77103-102EA 23 USD
77103-102 77103-104 77103-106
2-Bromo-5-hydroxymethylaniline 97%
2-Bromo-5-hydroxymethylaniline

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MDL Number: MFCD17214967
CAS Number: 1261666-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR