(4-((tert-Butoxycarbonyl)amino)-2-fluorophenyl)boronic acid 98%

Supplier: Ambeed
Warning

A511569-1G A511569-250MG A511569-5G
77103-090EA 113 USD
77103-090 77103-092 77103-094
(4-((tert-Butoxycarbonyl)amino)-2-fluorophenyl)boronic acid 98%
(4-((tert-Butoxycarbonyl)amino)-2-fluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BFNO₄
MDL Number: MFCD18250042
CAS Number: 1415960-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR