1-Amino-3,3-dimethyl-butan-2-one hydrochloride 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Amino-3,3-dimethylbutan-2-one hydrochloride
A509593-100G
A509593-10G
A509593-1G
A509593-250MG
A509593-25G
A509593-5G
77103-078EA
512.42
USD
77103-078
77103-080
77103-082
77103-084
77103-086
77103-088
1-Amino-3,3-dimethyl-butan-2-one hydrochloride 98%
1-Amino-3,3-dimethyl-butan-2-one hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₃NO·ClH MW: 151.634 g/mol |
MDL Number:
MFCD06798286 CAS Number: 33119-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...