tert-Butyl (2-(2-aminoethoxy)ethyl)carbamate hydrochloride 95%

Supplier: Ambeed
Warning

A508973-100MG A508973-1G A508973-250MG
77103-072EA 42.44 USD
77103-072 77103-074 77103-076
tert-Butyl (2-(2-aminoethoxy)ethyl)carbamate hydrochloride 95%
tert-Butyl (2-(2-aminoethoxy)ethyl)carbamate hydrochloride

Precaución: for Research Use Only

Formula: C₉H₂₁ClN₂O₃
MW: 240.72 g/mol
MDL Number: MFCD11844786
CAS Number: 165391-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR