2-Chlorothioanisole 97%

Supplier: Ambeed
Warning

Synonyms: 1-Chloro-2-(methylthio)benzene, 2-Chlorophenyl methyl sulphide

A507495-25G A507495-5G
77103-064EA 43.5 USD
77103-064 77103-066
2-Chlorothioanisole 97%
2-Chlorothioanisole

Precaución: for Research Use Only

Formula: C₇H₇ClS
MW: 158.65 g/mol
MDL Number: MFCD00040124
CAS Number: 17733-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR