4-(tert-Butyldimethylsilyloxy)-2,3-difluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-(t-Butyldimethylsilyloxy)-2,3-difluorophenylboronic acid

A507055-1G A507055-250MG A507055-5G
77103-052EA 106.09 USD
77103-052 77103-054 77103-056
4-(tert-Butyldimethylsilyloxy)-2,3-difluorophenylboronic acid 98%
4-(tert-Butyldimethylsilyloxy)-2,3-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₉BF₂O₃Si
MW: 288.171 g/mol
MDL Number: MFCD18914502
CAS Number: 870646-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR