2-Chloro-3-methyl-5-nitropyridine 98%

Supplier: Ambeed
Danger

Synonyms: 2-Chloro-5-nitro-3-picoline

A504868-10G A504868-1G A504868-25G A504868-5G A504868-100G
77103-044EA 9 USD
77103-044 77103-046 77103-048 77103-050 77613-590
2-Chloro-3-methyl-5-nitropyridine 98%
2-Chloro-3-methyl-5-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
MDL Number: MFCD00173686
CAS Number: 22280-56-4
UN: 2923
ADR: 8,6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR