Naringin dihydrochalcone 97%

Supplier: Ambeed
Warning

A504448-100G A504448-25G A504448-5G A504448-10G
77103-038EA 55 USD
77103-038 77103-040 77103-042 77901-665
Naringin dihydrochalcone 97%
Naringin dihydrochalcone

Precaución: for Research Use Only

Formula: C₂₇H₃₄O₁₄
MDL Number: MFCD08436145
CAS Number: 18916-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR