2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)malonic acid 96%

Supplier: Ambeed
Warning

A503145-100MG A503145-250MG A503145-5G A503145-1G
77103-016EA 42.44 USD
77103-016 77103-018 77103-020 77686-317
2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)malonic acid 96%
2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)malonic acid

Precaución: for Research Use Only

Formula: C₁₈H₁₅NO₆
MW: 341.32 g/mol
MDL Number: MFCD09841915
CAS Number: 296261-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR