3-(Cyclopentyloxy)-2,4-difluorophenylboronic acid 98%

Supplier: Ambeed
Warning

A502716-1G A502716-250MG A502716-5G
77103-004EA 133.68 USD
77103-004 77103-006 77103-008
3-(Cyclopentyloxy)-2,4-difluorophenylboronic acid 98%
3-(Cyclopentyloxy)-2,4-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃BF₂O₃
MW: 242.03 g/mol
MDL Number: MFCD19981516
CAS Number: 1629971-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR