1-Benzyl-3-iodo-7-azaindole 98+%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyl-3-iodo-1H-pyrrolo[2,3-b]pyridine

A501449-1G A501449-250MG A501449-5G
77102-996EA 43 USD
77102-996 77102-998 77103-000
1-Benzyl-3-iodo-7-azaindole 98+%
1-Benzyl-3-iodo-7-azaindole

Precaución: for Research Use Only

Formula: C₁₄H₁₁IN₂
MDL Number: MFCD09834873
CAS Number: 1093759-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR