2-Amino-4-phenylthiazole-5-carbonitrile 95%

Supplier: Ambeed
Warning

A501339-1G A501339-250MG A501339-5G
77102-994EA 89 USD
77102-994 77900-935 77901-055
2-Amino-4-phenylthiazole-5-carbonitrile 95%
2-Amino-4-phenylthiazole-5-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₇N₃S
MDL Number: MFCD22683032
CAS Number: 704870-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR