tert-Butyl (1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropyl)carbamate 98%
Supplier: Ambeed
|
Warning
|
A501330-100MG
A501330-1G
A501330-250MG
77102-988EA
34
USD
77102-988
77102-990
77102-992
tert-Butyl (1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropyl)carbamate 98%
tert-Butyl (1-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)cyclopropyl)carbamate
Precaución: for Research Use Only
Formula:
C₂₀H₃₀BNO₄ |
MDL Number:
MFCD21602848 CAS Number: 1313441-88-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...