2-Methyl-4,5-diphenylthiazole 98%

Supplier: Ambeed
Warning

A500746-1G A500746-250MG A500746-5G
77102-982EA 24.41 USD
77102-982 77102-984 77102-986
2-Methyl-4,5-diphenylthiazole 98%
2-Methyl-4,5-diphenylthiazole

Precaución: for Research Use Only

Formula: C₁₆H₁₃NS
MW: 251.35 g/mol
MDL Number: MFCD00186396
CAS Number: 3755-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR