3-Methoxy-N,N-dimethylpropanamide 98%
Supplier: Ambeed
|
Warning
|
A499897-100G
A499897-25G
A499897-500G
77102-972EA
63
USD
77102-972
77102-974
77102-976
3-Methoxy-N,N-dimethylpropanamide 98%
3-Methoxy-N,N-dimethylpropanamide
Precaución: for Research Use Only
Formula:
C₆H₁₃NO₂ |
MDL Number:
MFCD18970488 CAS Number: 53185-52-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...