5-(3-Methoxyphenyl)-1,3,4-thiadiazol-2-amine 97%

Supplier: Ambeed
Warning

A499649-1G A499649-250MG A499649-25G A499649-5G
77102-964EA 40 USD
77102-964 77102-966 77102-968 77901-907
5-(3-Methoxyphenyl)-1,3,4-thiadiazol-2-amine 97%
5-(3-Methoxyphenyl)-1,3,4-thiadiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₉N₃OS
MDL Number: MFCD00628650
CAS Number: 247109-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR