3-Nitro-5-(trifluoromethyl)phenol 98%

Supplier: Ambeed
Warning

A498759-5G A498759-1G
77102-960EA 49 USD
77102-960 77686-307
3-Nitro-5-(trifluoromethyl)phenol 98%
3-Nitro-5-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₄F₃NO₃
MDL Number: MFCD04973779
CAS Number: 349-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR