2-Chloro-5-fluoropyridin-4-ol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-fluoro-pyridin-4-ol

A496880-10G A496880-1G A496880-250MG A496880-25G A496880-5G
77102-952EA 401.03 USD
77102-952 77102-954 77102-956 77102-958 77686-303
2-Chloro-5-fluoropyridin-4-ol 97%
2-Chloro-5-fluoropyridin-4-ol

Precaución: for Research Use Only

Formula: C₅H₃ClFNO
MW: 147.535 g/mol
MDL Number: MFCD13189882
CAS Number: 1196153-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR