3-Methoxy-6-methylpyridin-2-amine 95%

Supplier: Ambeed
Warning

A496767-100MG A496767-1G A496767-250MG
77102-946EA 12 USD
77102-946 77102-948 77102-950
3-Methoxy-6-methylpyridin-2-amine 95%
3-Methoxy-6-methylpyridin-2-amine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
MDL Number: MFCD09878490
CAS Number: 478913-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR