1-(Chloroacetyl)pyrrolidine 98%

Supplier: Ambeed
Warning

A494830-1G A494830-25G A494830-5G
77102-934EA 17 USD
77102-934 77102-936 77102-938
1-(Chloroacetyl)pyrrolidine 98%
1-(Chloroacetyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₆H₁₀ClNO
MDL Number: MFCD01217237
CAS Number: 20266-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR