2,5-Dimethyl-1H-pyrrole-3,4-dicarboxaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 3,4-Diformyl-2,5-dimethylpyrrole, 2,5-Dimethylpyrrole-3,4-dicarbaldehyde, 2,5-Dimethyl-1H-pyrrole-3,4-dicarbaldehyde

A494416-100MG A494416-1G
77102-930EA 94.43 USD
77102-930 77102-932
2,5-Dimethyl-1H-pyrrole-3,4-dicarboxaldehyde 95%
2,5-Dimethyl-1H-pyrrole-3,4-dicarboxaldehyde

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.16 g/mol
MDL Number: MFCD02660558
CAS Number: 56139-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR