4'-Chloro-[1,1'-biphenyl]-3-carboxaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 4'-Chloro-3-formylbiphenyl, 3-(4-Chlorophenyl)benzaldehyde, 4'-Chloro-[1,1'-biphenyl]-3-carbaldehyde

A493910-100MG A493910-250MG A493910-1G
77102-926EA 37.14 USD
77102-926 77102-928 77613-556
4'-Chloro-[1,1'-biphenyl]-3-carboxaldehyde 98%
4'-Chloro-[1,1'-biphenyl]-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₉ClO
MW: 216.663 g/mol
MDL Number: MFCD03424622
CAS Number: 139502-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR