Methyl 6-bromo-2-fluoro-3-methylbenzoate 95%

Supplier: Ambeed
Warning

A493348-100MG A493348-1G A493348-250MG A493348-5G
77102-918EA 30.77 USD
77102-918 77102-920 77102-922 77102-924
Methyl 6-bromo-2-fluoro-3-methylbenzoate 95%
Methyl 6-bromo-2-fluoro-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₂
MW: 247.061 g/mol
MDL Number: MFCD24848718
CAS Number: 1437780-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR