(2R,5R)-tert-Butyl 5-(hydroxymethyl)-2-methylpiperazine-1-carboxylate 98%

Supplier: Ambeed
Warning

A493256-100MG A493256-250MG A493256-1G
77102-910EA 42.44 USD
77102-910 77102-912 77572-566
(2R,5R)-tert-Butyl 5-(hydroxymethyl)-2-methylpiperazine-1-carboxylate 98%
(2R,5R)-tert-Butyl 5-(hydroxymethyl)-2-methylpiperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₃
MW: 230.30 g/mol
MDL Number: MFCD28100885
CAS Number: 1403898-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR