1-(3-Bromo-2-chlorophenyl)ethanone 98%

Supplier: Ambeed
Warning

A491271-250MG A491271-5G
77102-906EA 20.16 USD
77102-906 77102-908
1-(3-Bromo-2-chlorophenyl)ethanone 98%
1-(3-Bromo-2-chlorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₆BrClO
MW: 233.48 g/mol
MDL Number: MFCD18390299
CAS Number: 161957-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR