1-Bromo-8-chlorodibenzo[b,d]furan 97%

Supplier: Ambeed
Warning

A491247-100MG A491247-1G A491247-250MG A491247-25G A491247-5G
77102-896EA 6.37 USD
77102-896 77102-898 77102-900 77102-902 77102-904
1-Bromo-8-chlorodibenzo[b,d]furan 97%
1-Bromo-8-chlorodibenzo[b,d]furan

Precaución: for Research Use Only

Formula: C₁₂H₆BrClO
MW: 281.53 g/mol
CAS Number: 2173554-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR