3,4-Difluoro-2-methylbenzaldehyde 97%

Supplier: Ambeed
Warning

A490012-1G A490012-250MG A490012-5G
77102-882EA 78 USD
77102-882 77102-884 77102-886
3,4-Difluoro-2-methylbenzaldehyde 97%
3,4-Difluoro-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆F₂O
MDL Number: MFCD07777151
CAS Number: 847502-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR