2,4-Dichloropyrimidine-5-carbonyl chloride 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
2,4-Dichloro-5-pyrimidinecarbonyl chloride
A489272-1G
A489272-250MG
77102-880EA
36
USD
77102-880
77686-278
2,4-Dichloropyrimidine-5-carbonyl chloride 95%
2,4-Dichloropyrimidine-5-carbonyl chloride
Precaución: for Research Use Only
Formula:
C₅HCl₃N₂O MW: 211.44 g/mol |
MDL Number:
MFCD09742432 CAS Number: 2972-52-3 UN: 3265 ADR: 8 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...