tert-Butyl (4-fluoropyridin-2-yl)carbamate 95%

Supplier: Ambeed
Warning

A489160-100MG A489160-250MG A489160-1G
77102-876EA 12 USD
77102-876 77102-878 77613-534
tert-Butyl (4-fluoropyridin-2-yl)carbamate 95%
tert-Butyl (4-fluoropyridin-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₃FN₂O₂
MDL Number: MFCD21362361
CAS Number: 1237535-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR