1-Amino-3-(dimethylamino)propan-2-ol 98%

Supplier: Ambeed
Danger

A488417-1G A488417-250MG A488417-100MG
77102-868EA 179 USD
77102-868 77102-870 77901-395
1-Amino-3-(dimethylamino)propan-2-ol 98%
1-Amino-3-(dimethylamino)propan-2-ol

Precaución: for Research Use Only

Formula: C₅H₁₄N₂O
MDL Number: MFCD09740503
CAS Number: 50411-39-7
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR