3-[(Dimethylamino)methyl]-4-hydroxy-5-methoxybenzenecarbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 3-((Dimethylamino)methyl)-4-hydroxy-5-methoxybenzaldehyde

A487148-10G A487148-1G A487148-250MG A487148-5G
77102-848EA 138.98 USD
77102-848 77102-850 77102-852 77102-854
3-[(Dimethylamino)methyl]-4-hydroxy-5-methoxybenzenecarbaldehyde 98%
3-[(Dimethylamino)methyl]-4-hydroxy-5-methoxybenzenecarbaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₃
MW: 209.25 g/mol
MDL Number: MFCD03011572
CAS Number: 87743-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR