4-(2,6-Dichlorobenzamido)-N-(piperidin-4-yl)-1H-pyrazole-3-carboxamide 98+%

Supplier: Ambeed
Warning

Synonyms: AT7519

A485554-10MG A485554-1MG A485554-50MG A485554-5MG
77102-834EA 55.17 USD
77102-834 77102-836 77102-838 77102-840
4-(2,6-Dichlorobenzamido)-N-(piperidin-4-yl)-1H-pyrazole-3-carboxamide 98+%
4-(2,6-Dichlorobenzamido)-N-(piperidin-4-yl)-1H-pyrazole-3-carboxamide

Precaución: for Research Use Only

Formula: C₁₆H₁₇Cl₂N₅O₂
MW: 382.24 g/mol
MDL Number: MFCD13184820
CAS Number: 844442-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR