2,4,5-Trifluorobenzylamine 98%

Supplier: Ambeed
Danger

A484272-250MG A484272-5G
77102-830EA 41.38 USD
77102-830 77686-275
2,4,5-Trifluorobenzylamine 98%
2,4,5-Trifluorobenzylamine

Precaución: for Research Use Only

Formula: C₇H₆F₃N
MW: 161.13 g/mol
MDL Number: MFCD00236316
CAS Number: 168644-93-7
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR