(4-Methylmorpholin-2-yl)methanamine dihydrochloride 95%

Supplier: Ambeed
Warning

A483799-100MG A483799-1G A483799-250MG
77102-824EA 25 USD
77102-824 77102-826 77102-828
(4-Methylmorpholin-2-yl)methanamine dihydrochloride 95%
(4-Methylmorpholin-2-yl)methanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₆Cl₂N₂O
MDL Number: MFCD21362291
CAS Number: 259090-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR