4-Bromo-5-chlorobenzene-1,2-diamine 95%

Supplier: Ambeed
Warning

A480917-100MG A480917-1G A480917-250MG
77102-814EA 35.01 USD
77102-814 77102-816 77102-818
4-Bromo-5-chlorobenzene-1,2-diamine 95%
4-Bromo-5-chlorobenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₆H₆BrClN₂
MW: 221.482 g/mol
MDL Number: MFCD16987754
CAS Number: 75293-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR