2,5-Dioxopyrrolidin-1-yl 4,7,10,13-tetraoxahexadec-15-yn-1-oate ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Propargyl-PEG4-NHS ester
A480509-1G
A480509-250MG
77102-802EA
132.62
USD
77102-802
77686-269
2,5-Dioxopyrrolidin-1-yl 4,7,10,13-tetraoxahexadec-15-yn-1-oate ≥98%
2,5-Dioxopyrrolidin-1-yl 4,7,10,13-tetraoxahexadec-15-yn-1-oate
Precaución: for Research Use Only
Formula:
C₁₆H₂₃NO₈ MW: 357.36 g/mol |
MDL Number:
MFCD22380743 CAS Number: 1428629-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...