Methyl 3-amino-5-fluoropicolinate 95%

Supplier: Ambeed
Warning

A479504-100MG A479504-1G A479504-250MG
77102-796EA 84 USD
77102-796 77102-798 77102-800
Methyl 3-amino-5-fluoropicolinate 95%
Methyl 3-amino-5-fluoropicolinate

Precaución: for Research Use Only

Formula: C₇H₇FN₂O₂
MW: 170.14 g/mol
MDL Number: MFCD18259055
CAS Number: 1052714-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR