(R)-tert-Butyl 7-methyl-1,4-diazepane-1-carboxylate 98%

Supplier: Ambeed
Warning

A477730-1G A477730-250MG
77102-784EA 187.78 USD
77102-784 77102-786
(R)-tert-Butyl 7-methyl-1,4-diazepane-1-carboxylate 98%
(R)-tert-Butyl 7-methyl-1,4-diazepane-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.30 g/mol
MDL Number: MFCD27986952
CAS Number: 1638743-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR