5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 5-Fluoro-2-(methoxycarbonyl)phenylboronic acid pinacol ester

A477659-100MG A477659-1G A477659-250MG A477659-5G
77102-776EA 16 USD
77102-776 77102-778 77102-780 77102-782
5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester 98%
5-Fluoro-2-(methoxycarbonyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BFO₄
MW: 280.10 g/mol
MDL Number: MFCD16996304
CAS Number: 1400976-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR