Sodium benzoate 98%

Supplier: Ambeed
Warning

Synonyms: Benzoic acid sodium salt

A477289-1000G A477289-100G A477289-25G A477289-500G
77102-768EA 19.1 USD
77102-768 77102-770 77102-772 77102-774
Sodium benzoate 98%
Sodium benzoate

Precaución: for Research Use Only

Formula: C₆H₅COONa
MW: 144.11 g/mol
MDL Number: MFCD00012463
CAS Number: 532-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR