6-Bromo-5-fluoropyridin-2-amine 98%

Supplier: Ambeed
Warning

A477187-100MG A477187-1G A477187-250MG A477187-5G
77102-762EA 40 USD
77102-762 77102-764 77102-766 77686-261
6-Bromo-5-fluoropyridin-2-amine 98%
6-Bromo-5-fluoropyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄BrFN₂
MDL Number: MFCD26407134
CAS Number: 1257294-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR