(2S,3R,4S,5S,6R)-6-((Trityloxy)methyl)tetrahydro-2H-pyran-2,3,4,5-tetraol 95%

Supplier: Ambeed
Warning

A476635-100MG A476635-250MG
77102-752EA 65.78 USD
77102-752 77102-754
(2S,3R,4S,5S,6R)-6-((Trityloxy)methyl)tetrahydro-2H-pyran-2,3,4,5-tetraol 95%
(2S,3R,4S,5S,6R)-6-((Trityloxy)methyl)tetrahydro-2H-pyran-2,3,4,5-tetraol

Precaución: for Research Use Only

Formula: C₂₅H₂₆O₆
MW: 422.47 g/mol
MDL Number: MFCD00061632
CAS Number: 54325-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR